2,3,4-Trimethoxybenzylamine

Supplier: Apollo Scientific
Danger

OR49012-5G OR49012-1G
APOSOR49012-5GEA 387 GBP
APOSOR49012-5G APOSOR49012-1G
2,3,4-Trimethoxybenzylamine
2,3,4-Trimethoxybenzylamine
Formula: C₁₀H₁₅NO₃
Storage Temperature: Ambient
CAS Number: 41219-16-3
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR