1-(1-Boc-3-azetidinyl)pyrrolidine 96%

Supplier: Apollo Scientific

OR480770-1G OR480770-250MG
APOSOR480770-1GEA 317 GBP
APOSOR480770-1G APOSOR480770-250MG
1-(1-Boc-3-azetidinyl)pyrrolidine 96%
1-(1-Boc-3-azetidinyl)pyrrolidine
Formula: C₁₂H₂₂N₂O₂
MW: 226,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09860616
CAS Number: 1019008-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR