3-(Bromomethyl)benzoic acid 95%

Supplier: Apollo Scientific

OR480743-25G OR480743-5G
APOSOR480743-25GEA 106 GBP
APOSOR480743-25G APOSOR480743-5G
3-(Bromomethyl)benzoic acid 95%
3-(Bromomethyl)benzoic acid
Formula: C8H7BrO2
MW: 215,0440 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045839
CAS Number: 6515-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR