5-Formylthiophene-2-carbonitrile ≥96%

Supplier: Apollo Scientific

Synonyms: 5-Cyanothiophene-2-carboxaldehyde

OR480690-1G OR480690-5G
APOSOR480690-1GEA 62.9 GBP
APOSOR480690-1G APOSOR480690-5G
5-Formylthiophene-2-carbonitrile ≥96%
5-Formylthiophene-2-carbonitrile
Formula: C₆H₃NOS
MW: 137,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037804
CAS Number: 21512-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR