1-Benzyl-3-pyrroline 96%

Supplier: Apollo Scientific

Synonyms: 1-Benzyl-2,5-dihydropyrrole

OR480683-25G OR480683-5G
APOSOR480683-25GEA 336 GBP
APOSOR480683-25G APOSOR480683-5G
1-Benzyl-3-pyrroline 96%
1-Benzyl-3-pyrroline
Formula: C₁₁H₁₃N
MW: 159,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012324
CAS Number: 6913-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR