Boc-Dap(Cbz)-OH 97%

Supplier: Apollo Scientific

Synonyms: Boc-Dap(Z)-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-3-(benzyloxycarbonylamino)propionic acid

OR480680-25G OR480680-5G
APOSOR480680-25GEA 106 GBP
APOSOR480680-25G APOSOR480680-5G
Boc-Dap(Cbz)-OH 97%
Boc-Dap(Cbz)-OH
Formula: C₁₆H₂₂N₂O₆
MW: 338,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275856
CAS Number: 65710-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR