Benzyl acrylate 97%

Supplier: Apollo Scientific

Synonyms: Acrylic Acid Benzyl Ester

OR480651-100G OR480651-250G
APOSOR480651-100GEA 154 GBP
APOSOR480651-100G APOSOR480651-250G
Benzyl acrylate 97%
Benzyl acrylate
Formula: C₁₀H₁₀O₂
MW: 162,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048147
CAS Number: 2495-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR