1-(4-Methylphenyl)cyclopropanecarboxylic acid 95%
Supplier: Apollo Scientific
Synonyms:
1-(p-Tolyl)cyclopropanecarboxylic acid
OR480650-10G
OR480650-5G
APOSOR480650-10GEA
215
GBP
APOSOR480650-10G
APOSOR480650-5G
1-(4-Methylphenyl)cyclopropanecarboxylic acid 95%
1-(4-Methylphenyl)cyclopropanecarboxylic acid
Formula:
C₁₁H₁₂O₂ MW: 176,2100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066918 CAS Number: 83846-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...