4-Methyl-3-nitrobenzenesulphonamide 97%
Supplier: Apollo Scientific
OR480648-10G
OR480648-2.5G
OR480648-500MG
APOSOR480648-10GEA
488
GBP
APOSOR480648-10G
APOSOR480648-2.5G
APOSOR480648-500MG
4-Methyl-3-nitrobenzenesulphonamide 97%
4-Methyl-3-nitrobenzenesulphonamide
Formula:
C₇H₈N₂O₄S MW: 216,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00460611 CAS Number: 6949-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...