N-(4-Oxocyclohexyl)acetamide 97%

Supplier: Apollo Scientific

Synonyms: 4-Acetamidocyclohexanone

OR480579-100G OR480579-250G
APOSOR480579-100GEA 109 GBP
APOSOR480579-100G APOSOR480579-250G
N-(4-Oxocyclohexyl)acetamide 97%
N-(4-Oxocyclohexyl)acetamide
Formula: C₈H₁₃NO₂
MW: 155,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03703462
CAS Number: 27514-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR