4-Isopropylbenzoic acid 95%

Supplier: Apollo Scientific

OR480571-100G OR480571-250G
APOSOR480571-100GEA 133 GBP
APOSOR480571-100G APOSOR480571-250G
4-Isopropylbenzoic acid 95%
4-Isopropylbenzoic acid
Formula: C₁₀H₁₂O₂
MW: 164,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002564
CAS Number: 536-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR