Methyl (4-methoxyphenyl)acetate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 4-methoxyphenylacetate

OR480569-100G OR480569-500G
APOSOR480569-100GEA 144 GBP
APOSOR480569-100G APOSOR480569-500G
Methyl (4-methoxyphenyl)acetate 97%
Methyl (4-methoxyphenyl)acetate
Formula: C₁₀H₁₂O₃
MW: 180,23 g/mol
Boiling Pt: 258 °C/19mmHg
Density: 1,117 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00008454
CAS Number: 23786-14-3
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR