4-(Styryl)pyrimidine 95%
Supplier: Apollo Scientific
OR480204-10G
OR480204-1G
OR480204-5G
APOSOR480204-10GEA
735
GBP
APOSOR480204-10G
APOSOR480204-1G
APOSOR480204-5G
4-(Styryl)pyrimidine 95%
4-(Styryl)pyrimidine
Formula:
C₁₂H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00188074 CAS Number: 39816-21-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...