4-Amino-2,6-dichlorophenol

Supplier: Apollo Scientific
Warning

OR4785-100G OR4785-25G
APOSOR4785-100GEA 27.2 GBP
APOSOR4785-100G APOSOR4785-25G
4-Amino-2,6-dichlorophenol
4-Amino-2,6-dichlorophenol
Formula: C₆H₅Cl₂NO
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007875
CAS Number: 5930-28-9
EINECS: 227-671-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR