Dithizone 80%

Supplier: Apollo Scientific
Warning

Synonyms: Dithizone,for analysis, 1,5-Diphenylthiocarbazone

OR47831-25G OR47831-5G
APOSOR47831-25GEA 38.1 GBP
APOSOR47831-25G APOSOR47831-5G
Dithizone 80%
Dithizone
Formula: C₆H₅NHNHCSN=NC₆H₅
MW: 256,33 g/mol
Melting Pt: 168 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003025
CAS Number: 60-10-6
EINECS: 200-454-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR