2-Chloro-3-methylquinoline 97%
Supplier: Apollo Scientific
OR471650-10G
OR471650-5G
APOSOR471650-10GEA
382
GBP
APOSOR471650-10G
APOSOR471650-5G
2-Chloro-3-methylquinoline 97%
2-Chloro-3-methylquinoline
Formula:
C₁₀H₈ClN MW: 177,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00100265 CAS Number: 57876-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...