(S)-β-(Carbobenzoxyamino)-γ-butyrolactone 98%

Supplier: Apollo Scientific

Synonyms: (S)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate, (S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (S)-beta-(Cbz-amino)-gamma-butyrolactone, (S)-4-(Cbz-amino)tetrahydrofuran-2-one

OR471646-10G OR471646-25G OR471646-5G
APOSOR471646-10GEA 138 GBP
APOSOR471646-10G APOSOR471646-25G APOSOR471646-5G
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone 98%
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone
Formula: C₁₂H₁₃NO₄
MW: 235,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00216569
CAS Number: 87219-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR