L-Tyrosine-tert-butyl ester 98%

Supplier: Apollo Scientific

Synonyms: H-Tyr-OtBu

OR471624-100G OR471624-25G
APOSOR471624-100GEA 231 GBP
APOSOR471624-100G APOSOR471624-25G
L-Tyrosine-tert-butyl ester 98%
L-Tyrosine-tert-butyl ester
Formula: C₁₃H₁₉NO₃
MW: 237,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042644
CAS Number: 16874-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR