3-m-Tolyloxypropylamine
Supplier: Apollo Scientific
Synonyms:
3-(3-Methylphenoxy)propylamine
OR470094-1G
OR470094-5G
APOSOR470094-1GEA
231
GBP
APOSOR470094-1G
APOSOR470094-5G
3-m-Tolyloxypropylamine
3-m-Tolyloxypropylamine
Formula:
C₁₀H₁₅NO MW: 165.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08271804 CAS Number: 26646-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...