Methyl-5-bromo-3-methylpicolinate 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester, Methyl 5-bromo-3-methylpyridine-2-carboxylate, Methyl 5-bromo-3-methylpicolinate

OR46522-1G OR46522-10G OR46522-5G
APOSOR46522-1GEA 85.2 GBP
APOSOR46522-1G APOSOR46522-10G APOSOR46522-5G
Methyl-5-bromo-3-methylpicolinate 98%
Methyl 5-bromo-3-methylpicolinate
Formula: C₈H₈BrNO₃
MW: 230,0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375126
CAS Number: 213771-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR