(S)-(+)-2-(Methoxymethyl)pyrrolidine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(S)-2-(Methoxymethyl)pyrrolidine
OR4615-1G
OR4615-250MG
APOSOR4615-1GEA
34.2
GBP
APOSOR4615-1G
APOSOR4615-250MG
(S)-(+)-2-(Methoxymethyl)pyrrolidine
(S)-(+)-2-(Methoxymethyl)pyrrolidine
Formula:
C₆H₁₃NO MW: 115,18 g/mol Boiling Pt: 61…62 °C/40mmHg Density: 0,932 g/cm³ (25 °C) Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010408 CAS Number: 63126-47-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...