2-Bromo-4-chlorobenzonitrile

Supplier: Apollo Scientific
Warning

OR46060-1G OR46060-5G
APOSOR46060-1GEA 47.2 GBP
APOSOR46060-1G APOSOR46060-5G
2-Bromo-4-chlorobenzonitrile
2-Bromo-4-chlorobenzonitrile
Formula: C₇H₃BrClN
MW: 216,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00158966
CAS Number: 57381-49-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR