2-Ethylhexyl bromide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane
OR460077-100G
OR460077-500G
APOSOR460077-100GEA
39.4
GBP
APOSOR460077-100G
APOSOR460077-500G
2-Ethylhexyl bromide
2-Ethylhexyl bromide
Formula:
C₈H₁₇Br MW: 193,13 g/mol Boiling Pt: 110…111 °C Density: 1,086 g/cm³ (25 °C) Flash Pt: 69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000220 CAS Number: 18908-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...