1,2,2-Trichloropropane 97%
Supplier: Apollo Scientific
OR460037-1G
OR460037-5G
APOSOR460037-1GEA
257
GBP
APOSOR460037-1G
APOSOR460037-5G
1,2,2-Trichloropropane 97%
1,2,2-Trichloropropane
Formula:
ClH₂CCCl₂CH₃ MW: 147,43 g/mol Boiling Pt: 121…123 °C Density: 1,3145 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS Number:
3175-23-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...