(S)-3-Hydroxypiperidine hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-3-Piperidinol hydrochloride

OR4577-5G OR4577-25G
APOSOR4577-5GEA 44.6 GBP
APOSOR4577-5G APOSOR4577-25G
(S)-3-Hydroxypiperidine hydrochloride 98%
(S)-3-Hydroxypiperidine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137,6100 g/mol
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD03426348
CAS Number: 475058-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR