Benzyl-4-oxo-1-piperidinecarboxylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Cbz-Piperidin-4-one, 1-Carbobenzoxy-4-piperidone, 1-Cbz-4-piperidone, 4-Oxo-1-piperidinecarboxylic Acid Benzyl Ester, 1-Benzyloxycarbonyl-4-piperidine

OR4552-100G OR4552-25G
APOSOR4552-100GEA 68.3 GBP
APOSOR4552-100G APOSOR4552-25G
Benzyl-4-oxo-1-piperidinecarboxylate 97%
Benzyl-4-oxo-1-piperidinecarboxylate
Formula: C₁₃H₁₅NO₃
MW: 233,27 g/mol
Boiling Pt: 114…140 °C/0,24mmHg
Melting Pt: 38…47 °C
Density: 1,172 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00673144
CAS Number: 19099-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR