2-(4-Bromophenoxy)propanoic acid 97%

Supplier: Apollo Scientific

OR452059-1G OR452059-5G
APOSOR452059-1GEA 183 GBP
APOSOR452059-1G APOSOR452059-5G
2-(4-Bromophenoxy)propanoic acid 97%
2-(4-Bromophenoxy)propanoic acid
Formula: C₉H₉BrO₃
MW: 245.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02295723
CAS Number: 32019-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR