3-(Boc-amino)-3-(2-methylphenyl)propionic acid 97%

Supplier: Apollo Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(o-tolyl)propanoic acid

OR452039-1G OR452039-5G
APOSOR452039-1GEA 197 GBP
APOSOR452039-1G APOSOR452039-5G
3-(Boc-amino)-3-(2-methylphenyl)propionic acid 97%
3-(Boc-amino)-3-(2-methylphenyl)propionic acid
Formula: C₁₅H₂₁NO₄
MW: 279,3315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02090696
CAS Number: 284493-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR