2-Iodo-4-nitrophenol 97%

Supplier: Apollo Scientific
Warning

OR45113-1G OR45113-250MG
APOSOR45113-1GEA 125 GBP
APOSOR45113-1G APOSOR45113-250MG
2-Iodo-4-nitrophenol 97%
2-Iodo-4-nitrophenol
Formula: C₆H₄INO₃
MW: 265,01 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01165664
CAS Number: 89487-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR