5-Chloro-2-iodoaniline
Supplier: Apollo Scientific
|
Danger
|
OR4504-100G
OR4504-25G
OR4504-5G
APOSOR4504-100GEA
134
GBP
APOSOR4504-100G
APOSOR4504-25G
APOSOR4504-5G
5-Chloro-2-iodoaniline
5-Chloro-2-iodoaniline
Formula:
C₆H₅ClIN MW: 253,47 g/mol Melting Pt: 40…44 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD06738960 CAS Number: 6828-35-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...