2-Oxo-2,3-dihydrobenzooxazole-6-sulphonyl chloride

Supplier: Apollo Scientific
Danger

Synonyms: 2-Oxo-2,3-dihydrobenzo[d]oxazole-6-sulphonyl chloride

OR45020-1G OR45020-5G
APOSOR45020-1GEA 98.3 GBP
APOSOR45020-1G APOSOR45020-5G
2-Oxo-2,3-dihydrobenzooxazole-6-sulphonyl chloride
2-Oxo-2,3-dihydrobenzooxazole-6-sulphonyl chloride
Formula: C₇H₄ClNO₄S
MW: 233,63 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD03001119
CAS Number: 27685-90-1
EINECS: 248-605-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR