2-Bromonicotinic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-3-pyridinecarboxylic acid, 2-Bromopyridine-3-carboxylic acid
OR4493-5G
OR4493-25G
APOSOR4493-5GEA
13.2
GBP
APOSOR4493-5G
APOSOR4493-25G
2-Bromonicotinic acid
2-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ MW: 202,01 g/mol Melting Pt: 200…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646068 CAS Number: 35905-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...