3-Amino-4-chlorobenzonitrile 98%

Supplier: Apollo Scientific
Warning

OR4469-25G OR4469-100G OR4469-5G
APOSOR4469-25GEA 135 GBP
APOSOR4469-25G APOSOR4469-100G APOSOR4469-5G
3-Amino-4-chlorobenzonitrile 98%
3-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152,58 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00017089
CAS Number: 53312-79-1
EINECS: 258-471-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR