4-Iodo-o-anisidine

Supplier: Apollo Scientific
Warning

Synonyms: 4-Iodo-2-methoxyaniline

OR43624-1G OR43624-5G
APOSOR43624-1GEA 72.2 GBP
APOSOR43624-1G APOSOR43624-5G
4-Iodo-o-anisidine
4-Iodo-o-anisidine
Formula: C₇H₈INO
MW: 249,490 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11044438
CAS Number: 338454-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR