2,3-Dichloro-5-(chloromethyl)pyridine

Supplier: Apollo Scientific
Warning

Synonyms: 5-(Chloromethyl)-2,3-dichloropyridine

OR43617-1G OR43617-500MG
APOSOR43617-1GEA 414 GBP
APOSOR43617-1G APOSOR43617-500MG
2,3-Dichloro-5-(chloromethyl)pyridine
2,3-Dichloro-5-(chloromethyl)pyridine
Formula: C₆H₄Cl₃N
MW: 196,46 g/mol
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00125365
CAS Number: 54127-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR