7-Bromo-4-chloroquinoline 98%

Supplier: Apollo Scientific

OR43544-1G OR43544-5G
APOSOR43544-1GEA 129 GBP
APOSOR43544-1G APOSOR43544-5G
7-Bromo-4-chloroquinoline 98%
7-Bromo-4-chloroquinoline
Formula: C₉H₅BrClN
MW: 242,4997 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684221
CAS Number: 75090-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR