2-(Benzyloxy)acetaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: (Benzyloxy)acetaldehyde, Benzyloxyacetaldehyde

OR43518-1G OR43518-5G
APOSOR43518-1GEA 111 GBP
APOSOR43518-1G APOSOR43518-5G
2-(Benzyloxy)acetaldehyde
2-(Benzyloxy)acetaldehyde
Formula: C₉H₁₀O₂
MW: 150,177 g/mol
Boiling Pt: 118…120 °C/13mmHg
Density: 1,069 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191779
CAS Number: 60656-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR