N-Acetyl-D-mannosamine monohydrate

Supplier: Apollo Scientific
Warning

OR42152-1G OR42152-5G OR42152-25G
APOSOR42152-1GEA 52.5 GBP
APOSOR42152-1G APOSOR42152-5G APOSOR42152-25G
N-Acetyl-D-mannosamine monohydrate
N-Acetyl-D-mannosamine monohydrate
Formula: C₈H₁₅NO₆·H₂O
MW: 239,24 g/mol (221,22 anhy)
Storage Temperature: Freezer
MDL Number: MFCD00136044
CAS Number: 3615-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR