9-Bromo-9-phenylfluorene

Supplier: Apollo Scientific
Danger

Synonyms: 9-Bromo-9-phenyl-9H-fluorene

OR4204-1G OR4204-25G OR4204-5G
APOSOR4204-1GEA 39.3 GBP
APOSOR4204-1G APOSOR4204-25G APOSOR4204-5G
9-Bromo-9-phenylfluorene
9-Bromo-9-phenylfluorene
Formula: C₁₉H₁₃Br
MW: 321.23 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00075522
CAS Number: 55135-66-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR