2-Ethylpyrrole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Ethyl-1H-pyrrole
OR42007-1G
OR42007-5G
APOSOR42007-1GEA
72.2
GBP
APOSOR42007-1G
APOSOR42007-5G
2-Ethylpyrrole 98%
2-Ethylpyrrole
Formula:
C₆H₉N MW: 95,15 g/mol Boiling Pt: 164…165 °C/754mmHg Density: 0,929 g/cm³ (25 °C) Flash Pt: 62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00060477 CAS Number: 1551-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...