2,3-Dihydro-1-benzofuran-5-ol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3-Dihydro-5-hydroxybenzo[b]furan, 2,3-Dihydrobenzofuran-5-ol
OR42003-1G
OR42003-250MG
APOSOR42003-1GEA
265
GBP
APOSOR42003-1G
APOSOR42003-250MG
2,3-Dihydro-1-benzofuran-5-ol 97%
2,3-Dihydro-1-benzofuran-5-ol
Formula:
C₈H₈O₂ MW: 136,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09029838 CAS Number: 40492-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...