3-(1-Piperazino)propionitrile 97%

Supplier: Apollo Scientific
Warning

OR4194-10G OR4194-2G
APOSOR4194-10GEA 166 GBP
APOSOR4194-10G APOSOR4194-2G
3-(1-Piperazino)propionitrile 97%
3-(1-Piperazino)propionitrile
Formula: C₇H₁₃N₃
MW: 139,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093566
CAS Number: 34064-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR