3-Pyridineboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid

OR4119-25G OR4119-5G OR4119-100G
APOSOR4119-25GEA 28.9 GBP
APOSOR4119-25G APOSOR4119-5G APOSOR4119-100G
3-Pyridineboronic acid 95%
3-Pyridineboronic acid

Note: NMR spectrum must be run in D2O.

Formula: C₅H₆BNO₂
MW: 122,92 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR