4-Phenyl-2-pyrrolidone 95%
Supplier: Apollo Scientific
OR41147-10G
OR41147-1G
OR41147-5G
APOSOR41147-10GEA
89.2
GBP
APOSOR41147-10G
APOSOR41147-1G
APOSOR41147-5G
4-Phenyl-2-pyrrolidone 95%
4-Phenyl-2-pyrrolidone
Formula:
C₁₀H₁₁NO MW: 161,21 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01687226 CAS Number: 1198-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...