7-Methyl-1,4-benzothiazin-3-one 99%

Supplier: Apollo Scientific
Warning

OR40782-1G OR40782-5G
APOSOR40782-1GEA 44.6 GBP
APOSOR40782-1G APOSOR40782-5G
7-Methyl-1,4-benzothiazin-3-one 99%
7-Methyl-1,4-benzothiazin-3-one
Formula: C₉H₉NOS
MW: 179,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02660690
CAS Number: 30322-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR