Diethanolamine 99%

Supplier: Apollo Scientific
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

OR40730-2.5KG OR40730-500G
APOSOR40730-2.5KGEA 119 GBP
APOSOR40730-2.5KG APOSOR40730-500G
Diethanolamine 99%
Diethanolamine
Formula: C₄H₁₁NO₂
MW: 105,14 g/mol
Boiling Pt: 268…270 °C
Melting Pt: 28…30 °C
Density: 1,097 g/cm³ (25 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR