2-Amino-5-methoxypyridine 97%

Supplier: Apollo Scientific
Warning

OR40515-5G OR40515-1G OR40515-25G
APOSOR40515-5GEA 119 GBP
APOSOR40515-5G APOSOR40515-1G APOSOR40515-25G
2-Amino-5-methoxypyridine 97%
2-Amino-5-methoxypyridine
Formula: C₆H₈N₂O
MW: 124,1405 g/mol
Boiling Pt: 128…130 °C/10mmHg
Storage Temperature: Freezer
MDL Number: MFCD07374873
CAS Number: 10167-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR