2-Bromo-6-iodobenzonitrile

Supplier: Apollo Scientific

OR400749-1G OR400749-5G
APOSOR400749-1GEA 69.5 GBP
APOSOR400749-1G APOSOR400749-5G
2-Bromo-6-iodobenzonitrile
2-Bromo-6-iodobenzonitrile
Formula: C₇H₃BrIN
MW: 307,9139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619725
CAS Number: 1245648-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR