4-Bromo-3-methylbenzyl alcohol, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Bromo-3-methylphenyl)methanol
OR400665-1G
OR400665-5G
APOSOR400665-1GEA
56.5
GBP
APOSOR400665-1G
APOSOR400665-5G
4-Bromo-3-methylbenzyl alcohol, Technical Grade
4-Bromo-3-methylbenzyl alcohol
Formula:
C₈H₉BrO MW: 201,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09264005 CAS Number: 149104-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...