3-Bromo-o-anisaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-2-methoxybenzaldehyde
OR400651-1G
OR400651-5G
APOSOR400651-1GEA
84
GBP
APOSOR400651-1G
APOSOR400651-5G
3-Bromo-o-anisaldehyde
3-Bromo-o-anisaldehyde
Formula:
C₈H₇BrO₂ MW: 215,0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834811 CAS Number: 88275-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...